Assay Details
| Gene Symbol: stk17b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200535.1 | NP_956829.1 | 1 - 2 | 132 | 65 | |
| GenBank mRNA | BC056298.1 | - | 1 - 2 | 132 | 65 | |
More Information
Set Membership: |
|

stk17b
Genomic Map