Assay Details
| Gene Symbol: gnb5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200746.1 | NP_957040.1 | 11 - 12 | 1065 | 102 | |
| GenBank mRNA | BC059544.1 | - | 11 - 12 | 1065 | 102 | |
More Information
Set Membership: |
|

gnb5b
Genomic Map