Assay Details
| Gene Symbol: gnb5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200746.1 | NP_957040.1 | 12 - 13 | 1228 | 60 | |
| GenBank mRNA | BC059544.1 | - | 12 - 13 | 1228 | 60 | |
More Information
Set Membership: |
|

gnb5b
Genomic Map