Assay Details
| Gene Symbol: panx1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200916.1 | NP_957210.1 | 440 | 105 | ||
| GenBank mRNA | BC048058.1 | - | 440 | 105 | ||
| BC165839.1 | - | 1 - 2 | 221 | 105 | ||
| EU332874.1 | - | 1 - 2 | 187 | 105 | ||
More Information
Set Membership: |
|

panx1a
Genomic Map