Assay Details
| Gene Symbol: ankrd10a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200980.1 | NP_957274.1 | 4 - 5 | 745 | 115 | |
| GenBank mRNA | BC044376.1 | - | 4 - 5 | 745 | 115 | |
| BC155115.1 | - | 4 - 5 | 823 | 115 | ||
More Information
Set Membership: |
|

ankrd10a
Genomic Map