Assay Details
| Gene Symbol: lipca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201022.1 | NP_957316.1 | 2 - 3 | 323 | 86 | |
| GenBank mRNA | BC053243.1 | - | 2 - 3 | 323 | 86 | |
| BC163963.1 | - | 3 - 4 | 298 | 86 | ||
More Information
Set Membership: |
|

lipca
Genomic Map