Assay Details
| Gene Symbol: lipca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201022.1 | NP_957316.1 | 3 - 4 | 499 | 130 | |
| GenBank mRNA | BC053243.1 | - | 3 - 4 | 499 | 130 | |
| BC163963.1 | - | 4 - 5 | 474 | 130 | ||
More Information
Set Membership: |
|

lipca
Genomic Map