Assay Details
| Gene Symbol: rpsa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201052.1 | NP_957346.1 | 2 - 3 | 221 | 80 | |
| GenBank mRNA | AY391434.1 | - | 2 - 3 | 80 | ||
| BC044504.1 | - | 2 - 3 | 221 | 80 | ||
| BC062859.1 | - | 2 - 3 | 80 | |||
| BC164084.1 | - | 1 - 2 | 170 | 80 | ||
More Information
Set Membership: |
|

rpsa
Genomic Map