Assay Details
| Gene Symbol: rpsa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201052.1 | NP_957346.1 | 5 - 6 | 719 | 86 | |
| GenBank mRNA | AY391434.1 | - | 5 - 6 | 700 | 86 | |
| BC044504.1 | - | 5 - 6 | 719 | 86 | ||
| BC062859.1 | - | 5 - 6 | 702 | 86 | ||
| BC164084.1 | - | 4 - 5 | 668 | 86 | ||
More Information
Set Membership: |
|

rpsa
Genomic Map