Assay Details
| Gene Symbol: sh3glb2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201119.1 | NP_957413.1 | 2 - 3 | 291 | 98 | |
| XM_005171809.4 | XP_005171866.1 | 2 - 3 | 335 | 98 | ||
| XM_021476045.1 | XP_021331720.1 | 2 - 3 | 337 | 98 | ||
| XM_021476047.1 | XP_021331722.1 | 2 - 3 | 335 | 98 | ||
| GenBank mRNA | BC047195.1 | - | 2 - 3 | 291 | 98 | |
More Information
Set Membership: |
|

sh3glb2b
Genomic Map