Assay Details
| Gene Symbol: sh3glb2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201119.1 | NP_957413.1 | 9 - 10 | 1104 | 66 | |
| XM_005171808.4 | XP_005171865.1 | 12 - 13 | 1245 | 66 | ||
| XM_005171809.4 | XP_005171866.1 | 11 - 12 | 1235 | 66 | ||
| XM_021476044.1 | XP_021331719.1 | 13 - 14 | 1293 | 66 | ||
| XM_021476045.1 | XP_021331720.1 | 12 - 13 | 1282 | 66 | ||
| XM_021476046.1 | XP_021331721.1 | 11 - 12 | 1222 | 66 | ||
| XM_021476047.1 | XP_021331722.1 | 10 - 11 | 1211 | 66 | ||
| XM_021476048.1 | XP_021331723.1 | 11 - 12 | 1182 | 66 | ||
| XM_021476049.1 | XP_021331724.1 | 10 - 11 | 1157 | 66 | ||
| GenBank mRNA | BC047195.1 | - | 9 - 10 | 1104 | 66 | |
More Information
Set Membership: |
|

sh3glb2b
Genomic Map