Assay Details
| Gene Symbol: spop | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201130.1 | NP_957424.1 | 3 - 4 | 623 | 59 | |
| GenBank mRNA | BC053276.1 | - | 3 - 4 | 623 | 59 | |
| BC152667.1 | - | 5 - 6 | 820 | 59 | ||
More Information
Set Membership: |
|

spop
Genomic Map