Assay Details
| Gene Symbol: hmbsa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201154.1 | NP_957448.1 | 4 - 5 | 253 | 71 | |
| XM_005171816.4 | XP_005171873.1 | 4 - 5 | 393 | 71 | ||
| GenBank mRNA | BC053268.1 | - | 4 - 5 | 253 | 71 | |
More Information
Set Membership: |
|

hmbsa
Genomic Map