Assay Details
| Gene Symbol: klf6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201461.2 | NP_958869.2 | 1 - 2 | 330 | 71 | |
| GenBank mRNA | BC048893.1 | - | 1 - 2 | 288 | 71 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

klf6a| Gene Symbol: klf6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201461.2 | NP_958869.2 | 1 - 2 | 330 | 71 | |
| GenBank mRNA | BC048893.1 | - | 1 - 2 | 288 | 71 | |
Set Membership: |
|