Assay Details
| Gene Symbol: cbx8a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205616.2 | NP_991179.2 | 2 - 3 | 165 | 91 | |
| GenBank mRNA | AY179330.1 | - | 2 - 3 | 160 | 91 | |
| AY398319.1 | - | 2 - 3 | 160 | 91 | ||
| BC125904.1 | - | 2 - 3 | 165 | 91 | ||
More Information
Set Membership: |
|

cbx8a
Genomic Map