Assay Details
| Gene Symbol: slc31a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001320405.1 | NP_001307334.1 | 4 - 5 | 555 | 78 | |
| NM_205717.3 | NP_991280.2 | 4 - 5 | 545 | 78 | ||
| GenBank mRNA | AY077715.1 | - | 4 - 5 | 545 | 78 | |
| BC050484.1 | - | 4 - 5 | 546 | 78 | ||
| BC067161.1 | - | 4 - 5 | 495 | 78 | ||
| BC152191.1 | - | 4 - 5 | 559 | 78 | ||
| CF996108.1 | - | 4 - 5 | 500 | 78 | ||
More Information
Set Membership: |
|

slc31a1
Genomic Map