Assay Details
| Gene Symbol: fam234b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_207053.1 | NP_996936.1 | 8 - 9 | 1354 | 63 | |
| XM_005171506.4 | XP_005171563.1 | 8 - 9 | 1434 | 63 | ||
| GenBank mRNA | BC066400.1 | - | 8 - 9 | 1354 | 63 | |
More Information
Set Membership: |
|

fam234b
Genomic Map