Assay Details
| Gene Symbol: ewsr1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212630.1 | NP_997795.1 | 10 - 11 | 1052 | 123 | |
| XM_005165107.2 | XP_005165164.2 | 10 - 11 | 1039 | 123 | ||
| XM_005165108.2 | XP_005165165.2 | 10 - 11 | 1011 | 123 | ||
| XM_009301286.1 | XP_009299561.1 | 9 - 10 | 1007 | 123 | ||
| XM_009301287.1 | XP_009299562.1 | 9 - 10 | 1001 | 123 | ||
| GenBank mRNA | BC044518.1 | - | 10 - 11 | 1052 | 123 | |
| BC067661.1 | - | 9 - 10 | 1006 | 123 | ||
| BC097019.1 | - | 10 - 11 | 1045 | 123 | ||
| Gene Symbol: LOC110438494 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021471017.1 | XP_021326692.1 | 3 - 4 | 318 | 123 | |
| Gene Symbol: LOC110440168 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021480686.1 | XP_021336361.1 | 13 - 14 | 1440 | 123 | |
| XM_021480687.1 | XP_021336362.1 | 13 - 14 | 1434 | 123 | ||
| XM_021480688.1 | XP_021336363.1 | 13 - 14 | 1413 | 123 | ||
| XM_021480689.1 | XP_021336364.1 | 12 - 13 | 1281 | 123 | ||
| XM_021480691.1 | XP_021336366.1 | 12 - 13 | 1319 | 123 | ||
| XM_021480692.1 | XP_021336367.1 | 12 - 13 | 1215 | 123 | ||
| XM_021480693.1 | XP_021336368.1 | 12 - 13 | 1188 | 123 | ||
| XM_021480694.1 | XP_021336369.1 | 10 - 11 | 1017 | 123 | ||
| XM_021480695.1 | XP_021336370.1 | 10 - 11 | 1011 | 123 | ||
| XM_021480696.1 | XP_021336371.1 | 7 - 8 | 606 | 123 | ||
| Gene Symbol: clpxb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AI545751.1 | - | 1 - 2 | 106 | 123 | |
More Information
Set Membership: |
|

ewsr1b
Genomic Map