Assay Details
| Gene Symbol: ewsr1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212630.1 | NP_997795.1 | 12 - 13 | 1295 | 110 | |
| XM_005165107.2 | XP_005165164.2 | 12 - 13 | 1282 | 110 | ||
| XM_005165108.2 | XP_005165165.2 | 12 - 13 | 1254 | 110 | ||
| XM_009301286.1 | XP_009299561.1 | 11 - 12 | 1250 | 110 | ||
| XM_009301287.1 | XP_009299562.1 | 11 - 12 | 1244 | 110 | ||
| GenBank mRNA | BC044518.1 | - | 12 - 13 | 1295 | 110 | |
| BC067661.1 | - | 11 - 12 | 1249 | 110 | ||
| BC097019.1 | - | 12 - 13 | 1288 | 110 | ||
| Gene Symbol: LOC110438494 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021471017.1 | XP_021326692.1 | 5 - 6 | 561 | 110 | |
| Gene Symbol: LOC110440168 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021480686.1 | XP_021336361.1 | 15 - 16 | 1683 | 110 | |
| XM_021480687.1 | XP_021336362.1 | 15 - 16 | 1677 | 110 | ||
| XM_021480688.1 | XP_021336363.1 | 15 - 16 | 1656 | 110 | ||
| XM_021480689.1 | XP_021336364.1 | 14 - 15 | 1524 | 110 | ||
| XM_021480691.1 | XP_021336366.1 | 14 - 15 | 1562 | 110 | ||
| XM_021480692.1 | XP_021336367.1 | 14 - 15 | 1458 | 110 | ||
| XM_021480693.1 | XP_021336368.1 | 14 - 15 | 1431 | 110 | ||
| XM_021480694.1 | XP_021336369.1 | 12 - 13 | 1260 | 110 | ||
| XM_021480695.1 | XP_021336370.1 | 12 - 13 | 1254 | 110 | ||
| XM_021480696.1 | XP_021336371.1 | 9 - 10 | 849 | 110 | ||
| Gene Symbol: clpxb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AI545751.1 | - | 3 - 4 | 349 | 110 | |
More Information
Set Membership: |
|

ewsr1b
Genomic Map