Assay Details
| Gene Symbol: mbd3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212769.1 | NP_997934.1 | 1 - 2 | 235 | 82 | |
| GenBank mRNA | AY398379.1 | - | 1 - 2 | 235 | 82 | |
| BC071531.1 | - | 1 - 2 | 209 | 82 | ||
More Information
Set Membership: |
|

mbd3a
Genomic Map