Assay Details
| Gene Symbol: mbd3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212769.1 | NP_997934.1 | 5 - 6 | 792 | 73 | |
| XM_005168451.4 | XP_005168508.1 | 5 - 6 | 699 | 73 | ||
| GenBank mRNA | AY238337.1 | - | 5 - 6 | 651 | 73 | |
| AY398379.1 | - | 5 - 6 | 792 | 73 | ||
| BC071531.1 | - | 5 - 6 | 766 | 73 | ||
More Information
Set Membership: |
|

mbd3a
Genomic Map