Assay Details
| Gene Symbol: smc1al | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212810.2 | NP_997975.2 | 2 - 3 | 591 | 84 | |
| GenBank mRNA | AY648730.1 | - | 2 - 3 | 591 | 84 | |
| BC055212.1 | - | 2 - 3 | 584 | 84 | ||
| BC066674.1 | - | 2 - 3 | 513 | 84 | ||
More Information
Set Membership: |
|

smc1al
Genomic Map