Assay Details
| Gene Symbol: smc1al | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212810.2 | NP_997975.2 | 3 - 4 | 698 | 65 | |
| GenBank mRNA | AY648730.1 | - | 3 - 4 | 698 | 65 | |
| BC066674.1 | - | 3 - 4 | 620 | 65 | ||
More Information
Set Membership: |
|

smc1al
Genomic Map