Assay Details
| Gene Symbol: sstr5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213297.1 | NP_998462.1 | 2 - 3 | 1456 | 115 | |
| GenBank mRNA | BC068356.1 | - | 2 - 3 | 1456 | 115 | |
More Information
Set Membership: |
|

sstr5
Genomic Map