Assay Details
| Gene Symbol: decr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213321.1 | NP_998486.1 | 7 - 8 | 679 | 65 | |
| XM_005162880.4 | XP_005162937.1 | 7 - 8 | 709 | 65 | ||
| GenBank mRNA | BC068332.1 | - | 7 - 8 | 679 | 65 | |
More Information
Set Membership: |
|

decr2
Genomic Map