Assay Details
| Gene Symbol: cnn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213349.1 | NP_998514.1 | 2 - 3 | 212 | 81 | |
| XM_021479618.1 | XP_021335293.1 | 2 - 3 | 337 | 81 | ||
| GenBank mRNA | BC059802.1 | - | 2 - 3 | 212 | 81 | |
| DQ104245.1 | - | 2 - 3 | 218 | 81 | ||
More Information
Set Membership: |
|

cnn2
Genomic Map