Assay Details
| Gene Symbol: ndufa4l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213417.2 | NP_998582.1 | 3 - 4 | 320 | 119 | |
| GenBank mRNA | BC051617.1 | - | 3 - 4 | 292 | 119 | |
| EH577327.1 | - | 1 - 2 | 311 | 119 | ||
| EH585665.1 | - | 3 - 4 | 349 | 119 | ||
More Information
Set Membership: |
|

ndufa4l
Genomic Map