Assay Details
| Gene Symbol: creb3l3l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213532.2 | NP_998697.2 | 7 - 8 | 878 | 88 | |
| XM_005167154.3 | XP_005167211.1 | 7 - 8 | 1033 | 88 | ||
| GenBank mRNA | BC055664.1 | - | 7 - 8 | 926 | 88 | |
More Information
Set Membership: |
|

creb3l3l
Genomic Map