Assay Details
| Gene Symbol: nkx1.2lb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213548.1 | NP_998713.1 | 1 - 2 | 577 | 95 | |
| GenBank mRNA | AB121758.1 | - | 1 - 2 | 577 | 95 | |
| BC129149.1 | - | 1 - 2 | 578 | 95 | ||
More Information
Set Membership: |
|

nkx1.2lb
Genomic Map