Assay Details
| Gene Symbol: mtrex | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214768.1 | NP_999933.1 | 5 - 6 | 501 | 104 | |
| GenBank mRNA | BC061456.1 | - | 5 - 6 | 501 | 104 | |
More Information
Set Membership: |
|

mtrex
Genomic Map