Assay Details
| Gene Symbol: rela | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001839.2 | NP_001001839.2 | 10 - 11 | 1111 | 59 | |
| XM_005170796.4 | XP_005170853.1 | 10 - 11 | 1189 | 59 | ||
| GenBank mRNA | AY163839.1 | - | 10 - 11 | 1089 | 59 | |
| BC162910.1 | - | 10 - 11 | 1084 | 59 | ||
| BC162911.1 | - | 10 - 11 | 1084 | 59 | ||
More Information
Set Membership: |
|

rela
Genomic Map