Assay Details
| Gene Symbol: sf3b6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002067.1 | NP_001002067.1 | 2 - 3 | 301 | 62 | |
| GenBank mRNA | BC071385.1 | - | 2 - 3 | 301 | 62 | |
More Information
Set Membership: |
|

sf3b6
Genomic Map