Assay Details
| Gene Symbol: lsm6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002077.2 | NP_001002077.1 | 2 - 3 | 193 | 80 | |
| XM_005159968.4 | XP_005160025.1 | 2 - 3 | 435 | 80 | ||
| GenBank mRNA | BC072549.1 | - | 2 - 3 | 126 | 80 | |
| BC164231.1 | - | 128 | 80 | |||
| EH577307.1 | - | 2 - 3 | 335 | 80 | ||
| EH593457.1 | - | 2 - 3 | 179 | 80 | ||
More Information
Set Membership: |
|

lsm6
Genomic Map