Assay Details
| Gene Symbol: lsm6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002077.2 | NP_001002077.1 | 3 - 4 | 301 | 110 | |
| XM_005159968.4 | XP_005160025.1 | 3 - 4 | 543 | 110 | ||
| GenBank mRNA | BC072549.1 | - | 3 - 4 | 234 | 110 | |
| EH577307.1 | - | 1 - 2 | 227 | 110 | ||
More Information
Set Membership: |
|

lsm6
Genomic Map