Assay Details
| Gene Symbol: impdh1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002177.1 | NP_001002177.1 | 4 - 5 | 449 | 78 | |
| XM_005159007.4 | XP_005159064.1 | 4 - 5 | 995 | 78 | ||
| XM_005159008.4 | XP_005159065.1 | 4 - 5 | 997 | 78 | ||
| XM_009293441.2 | XP_009291716.1 | 4 - 5 | 484 | 78 | ||
| GenBank mRNA | BC074090.1 | - | 4 - 5 | 449 | 78 | |
| BC165226.1 | - | 3 - 4 | 279 | 78 | ||
More Information
Set Membership: |
|

impdh1a
Genomic Map