Assay Details
| Gene Symbol: impdh1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002177.1 | NP_001002177.1 | 8 - 9 | 1024 | 118 | |
| XM_005159007.4 | XP_005159064.1 | 8 - 9 | 1570 | 118 | ||
| XM_005159008.4 | XP_005159065.1 | 8 - 9 | 1572 | 118 | ||
| XM_009293441.2 | XP_009291716.1 | 8 - 9 | 1059 | 118 | ||
| GenBank mRNA | BC074090.1 | - | 8 - 9 | 1024 | 118 | |
| BC165226.1 | - | 7 - 8 | 854 | 118 | ||
More Information
Set Membership: |
|

impdh1a
Genomic Map