Assay Details
| Gene Symbol: nceh1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002303.1 | NP_001002303.1 | 3 - 4 | 468 | 90 | |
| GenBank mRNA | BC074087.1 | - | 3 - 4 | 468 | 90 | |
More Information
Set Membership: |
|

nceh1a
Genomic Map