Assay Details
| Gene Symbol: ufd1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002451.1 | NP_001002451.1 | 4 - 5 | 398 | 68 | |
| GenBank mRNA | AY648824.1 | - | 4 - 5 | 354 | 68 | |
| BC076020.1 | - | 4 - 5 | 398 | 68 | ||
| BC153586.1 | - | 4 - 5 | 328 | 68 | ||
More Information
Set Membership: |
|

ufd1l
Genomic Map