Assay Details
| Gene Symbol: brap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002466.1 | NP_001002466.1 | 6 - 7 | 1088 | 69 | |
| XM_021479154.1 | XP_021334829.1 | 6 - 7 | 1059 | 69 | ||
| XM_021479155.1 | XP_021334830.1 | 3 - 4 | 290 | 69 | ||
| GenBank mRNA | BC076350.1 | - | 6 - 7 | 1088 | 69 | |
| BC165196.1 | - | 6 - 7 | 886 | 69 | ||
More Information
Set Membership: |
|

brap
Genomic Map