Assay Details
| Gene Symbol: syn2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002597.2 | NP_001002597.2 | 6 - 7 | 928 | 81 | |
| GenBank mRNA | BC075998.1 | - | 6 - 7 | 928 | 81 | |
More Information
Set Membership: |
|

syn2a
Genomic Map