Assay Details
| Gene Symbol: syn2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002597.2 | NP_001002597.2 | 7 - 8 | 1071 | 108 | |
| GenBank mRNA | BC075998.1 | - | 7 - 8 | 1071 | 108 | |
More Information
Set Membership: |
|

syn2a
Genomic Map