Assay Details
| Gene Symbol: srsf5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002610.1 | NP_001002610.1 | 2 - 3 | 178 | 69 | |
| XM_005160668.4 | XP_005160725.1 | 2 - 3 | 283 | 69 | ||
| GenBank mRNA | BC075982.1 | - | 2 - 3 | 178 | 69 | |
| BC097003.1 | - | 2 - 3 | 206 | 69 | ||
More Information
Set Membership: |
|

srsf5b
Genomic Map