Assay Details
| Gene Symbol: srsf5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002610.1 | NP_001002610.1 | 5 - 6 | 432 | 80 | |
| GenBank mRNA | BC075982.1 | - | 5 - 6 | 432 | 80 | |
More Information
Set Membership: |
|

srsf5b
Genomic Map