Assay Details
| Gene Symbol: pdlim5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002654.2 | NP_001002654.2 | 8 - 9 | 1109 | 65 | |
| XM_017356059.2 | XP_017211548.1 | 9 - 10 | 1381 | 65 | ||
| XM_017356060.2 | XP_017211549.1 | 9 - 10 | 1172 | 65 | ||
| XM_021476075.1 | XP_021331750.1 | 10 - 11 | 917 | 65 | ||
| XM_021476076.1 | XP_021331751.1 | 10 - 11 | 914 | 65 | ||
| XM_021476078.1 | XP_021331753.1 | 9 - 10 | 899 | 65 | ||
| XM_021476079.1 | XP_021331754.1 | 9 - 10 | 896 | 65 | ||
| GenBank mRNA | BC076551.1 | - | 8 - 9 | 1109 | 65 | |
| BC165217.1 | - | 7 - 8 | 1010 | 65 | ||
More Information
Set Membership: |
|

pdlim5a
Genomic Map