Assay Details
| Gene Symbol: tomm20b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002698.1 | NP_001002698.1 | 4 - 5 | 504 | 59 | |
| GenBank mRNA | BC076116.1 | - | 4 - 5 | 504 | 59 | |
More Information
Set Membership: |
|

tomm20b
Genomic Map