Assay Details
| Gene Symbol: tspan18a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002734.2 | NP_001002734.1 | 8 - 9 | 958 | 73 | |
| XM_005172271.4 | XP_005172328.1 | 8 - 9 | 1008 | 73 | ||
| XM_005172272.4 | XP_005172329.1 | 8 - 9 | 999 | 73 | ||
| XM_005172273.4 | XP_005172330.1 | 7 - 8 | 933 | 73 | ||
| XM_009303047.3 | XP_009301322.1 | 9 - 10 | 1079 | 73 | ||
| XM_009303049.3 | XP_009301324.1 | 9 - 10 | 1070 | 73 | ||
| XM_021477461.1 | XP_021333136.1 | 7 - 8 | 1242 | 73 | ||
| XM_021477462.1 | XP_021333137.1 | 8 - 9 | 901 | 73 | ||
| XM_021477463.1 | XP_021333138.1 | 8 - 9 | 896 | 73 | ||
| GenBank mRNA | BC076426.1 | - | 8 - 9 | 962 | 73 | |
More Information
Set Membership: |
|

tspan18a
Genomic Map