Assay Details
| Gene Symbol: med19b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003416.2 | NP_001003416.2 | 3 - 4 | 559 | 72 | |
| GenBank mRNA | AY648741.1 | - | 3 - 4 | 465 | 72 | |
| BC092743.1 | - | 3 - 4 | 559 | 72 | ||
| BC165088.1 | - | 3 - 4 | 499 | 72 | ||
More Information
Set Membership: |
|

med19b
Genomic Map