Assay Details
| Gene Symbol: med19b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003416.2 | NP_001003416.2 | 4 - 5 | 658 | 156 | |
| GenBank mRNA | AY648741.1 | - | 4 - 5 | 564 | 156 | |
| BC092743.1 | - | 4 - 5 | 658 | 156 | ||
| BC165088.1 | - | 4 - 5 | 598 | 156 | ||
More Information
Set Membership: |
|

med19b
Genomic Map