Assay Details
| Gene Symbol: sfxn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003537.2 | NP_001003537.1 | 4 - 5 | 528 | 61 | |
| XM_005169627.4 | XP_005169684.1 | 5 - 6 | 605 | 61 | ||
| GenBank mRNA | BC078297.1 | - | 4 - 5 | 532 | 61 | |
More Information
Set Membership: |
|

sfxn1
Genomic Map