Assay Details
| Gene Symbol: dimt1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003556.3 | NP_001003556.2 | 1 - 2 | 165 | 118 | |
| GenBank mRNA | BC078286.1 | - | 1 - 2 | 159 | 118 | |
More Information
Set Membership: |
|

dimt1l
Genomic Map